Anti-KISS1R Rabbit Polyclonal Antibody

Supplier: Proteintech
15505-1-AP-150UL
10089-290EA 402 USD
10089-290
Anti-KISS1R Rabbit Polyclonal Antibody
Antibodies
receptor for metastin (kisspeptin-54, 13, 14, 10), C- terminally amidated peptides of KiSS-1. KiSS-1 is a metastasis suppressor protein that suppresses metastases in malignant melanomas and in some other carcinomas without affecting tumorigenicity. The metastasis suppressor properties may be mediated in part by cell cycle arrest and induction of apoptosis in malignant cells. The receptor is also probably involved in the regulation and fine-tuning of trophoblast invasion generated by the trophoblast itself. Analysis of the transduction pathways activated by the receptor identifies coupling to phospholipase C and intracellular calcium release through pertussis toxin-insensitive G(q) proteins.

Western Blot:SH-SY5Y Cells, 1:500-1:5000; IF: MCF-7, 1:10-1:100

Type: Primary
Antigen: KISS1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR