Anti-transgelin/SM22-specific Rabbit Polyclonal Antibody

Supplier: Proteintech
15502-1-AP-150UL
10095-584EA 402 USD
10095-584
Anti-transgelin/SM22-specific Rabbit Polyclonal Antibody
Antibodies
The transgelin family is a group of proteins that belong to 22kd actin-related corpnin superfamily. Of all three isoforms, transgelin 1 is the best characterized. Transgelin 1, also known as SM22alpha, is a specific marker for differentiated smooth muscle cells. Transgelin 2, also known as SM22 beta, is expressed by both smooth muscle and non-smooth muscle cells in a temporally and spatially regulated pattern. Trangenlin 3, also known as NP25, is only found in highly differentiated neuronal cells. This antibody is speific to transgelin 1. It does not cross-react with transgelin 2 and 3.

Western Blot: Human Colon Tissue, 1:200-1:2000

Type: Primary
Antigen: transgelin/SM22-specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR