Anti-MARK2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15492-1-AP-150UL
10089-986EA 402 USD
10089-986
Anti-MARK2 Rabbit Polyclonal Antibody
Antibodies
MARK2 is also named as EMK1(ELKL motif kinase 1), Par1b and belongs to the CAMK Ser/Thr protein kinase family. Par1b/MARK2 plays a critical role in neuronal polarity in the process of axon specification from multiple candidate neurites using primary cultures of mammalian hippocampal neurons. It has 16 isoforms produced by alternative promoter usage and alternative splicing and can exsit as a dimer.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: MARK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR