Anti-OSBPL10 Rabbit Polyclonal Antibody

Supplier: Proteintech
15491-1-AP-150UL
10091-620EA 402 USD
10091-620
Anti-OSBPL10 Rabbit Polyclonal Antibody
Antibodies
OSBPL10, also known as ORP10, is a member of the oxysterol-binding protein family. OSBPL10 localizes on microtubules and acts as a regulator of cellular lipid biosyntheses and apolipoprotein B-100. Genetic variation in OSBPL10 is associated with dyslipidmias and peripheral artery disease.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: OSBPL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR