Anti-ALKBH7 Rabbit Polyclonal Antibody

Supplier: Proteintech
15470-1-AP-150UL
10082-800EA 402 USD
10082-800
Anti-ALKBH7 Rabbit Polyclonal Antibody
Antibodies
ALKBH7(Alkylated DNA repair protein alkB homolog 7) is also named as ABH7, SPATA11 and belongs to the alkB family. This protein may be a dioxygenase that requires molecular oxygen, alpha-ketoglutarate and iron. Quantitative real-time PCR detectes variable ALKBH7 expression in all 16 tissues examined, with highest expression in pancreas, followed by spleen, prostate, ovary, and placenta. The gene encodes a 25 kDa protein with a 20 amino acid signal peptide.

Western Blot: Human Testis Tissue, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: ALKBH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR