Anti-CYB5B Rabbit Polyclonal Antibody

Supplier: Proteintech
15469-1-AP-150UL
10085-448EA 402 USD
10085-448
Anti-CYB5B Rabbit Polyclonal Antibody
Antibodies
CYB5B(Cytochrome b5 type B) is also named as CYB5M, OMB5 and belongs to the cytochrome b5 family. It is a 21 kDa membrane bound hemoprotein which function as an electron carrier for several membrane bound oxygenases. This protein is expressed on the plasma membrane of lymphoma cells but not normal lymphocytes, reactive lymphocytes or bone marrow precursor cells. The full length protein has a propeptide with 11 amino acids.

Western Blot: Mouse lung Tissue, 1:1000-1:10000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: CYB5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR