Anti-HIP14; ZDHHC17 Rabbit Polyclonal Antibody

Supplier: Proteintech
15465-1-AP-150UL
10088-256EA 402 USD
10088-256
Anti-HIP14; ZDHHC17 Rabbit Polyclonal Antibody
Antibodies
Post-translational modification by the lipid palmitate is crucial for the correct targeting and function of many proteins. Polyglutamine expansions of huntingtin protein are responsible for the Huntington neurological disorder. ZDHHC17 gene encodes palmitoyl transferase huntingtin interacting protein (HIP14) which regulates palmitoylation and distribution of hungtingtin. The latter is implicated in the formation of inclusion bodies and enhanced neuronal toxicity. HIP14 was also shown to control neurotransmitter release.

Western Blot: Raji Cells, 1:200-1:2000; IHC: Human Testis, 1:20-1:200

Type: Primary
Antigen: HIP14; ZDHHC17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR