Anti-RPP25 Rabbit Polyclonal Antibody

Supplier: Proteintech
15461-1-AP-150UL
10093-928EA 402 USD
10093-928
Anti-RPP25 Rabbit Polyclonal Antibody
Antibodies
Ribonuclease P protein subunit p25(RPP25) is a component of ribonuclease P, which is a protein complex that generates mature tRNA molecular by cleaving their 5'-ends. It belongs to histone-like Alba family, also a component of RNase MRP. This subunit binds to RNA.

Western Blot: HeLa Cells, 1:200-1:1000; IHC: Human small intestine Tissue, 1:20-1:200

Type: Primary
Antigen: RPP25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR