Anti-PAF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15441-1-AP-150UL
10091-734EA 524.48 USD
10091-734
Anti-PAF1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200; IP: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: PAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR