Anti-CA11 Rabbit Polyclonal Antibody

Supplier: Proteintech
15435-1-AP-150UL
10083-908EA 402 USD
10083-908
Anti-CA11 Rabbit Polyclonal Antibody
Antibodies
CA11(Carbonic anhydrase-related protein 11) is also named as CARP2, CARP XI and belongs to the alpha-carbonic anhydrase family. CARP XI is observed in the cerebellum, cerebrum, liver, stomach, small intestine, colon, kidney, and testis by immunohistochemical and in the cerebellum, the most prominent signal is located in the Purkinje cells. CARP XI contains a hydrophobic N-terminal region for a possible signal sequence and asparagine glycosylation site and it plays a general role in the human central nervous system.

Western Blot: Human Kidney Tissue, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: CA11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR