Anti-CD300LG Rabbit Polyclonal Antibody

Supplier: Proteintech
15426-1-AP-150UL
10084-322EA 402 USD
10084-322
Anti-CD300LG Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Kidney Tissue, 1:500-1:5000

Type: Primary
Antigen: CD300LG
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR