Anti-ECE2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15418-1-AP-150UL
10086-238EA 402 USD
10086-238
Anti-ECE2 Rabbit Polyclonal Antibody
Antibodies
Endothelin-converting enzymes (ECEs) are the key enzymes in the endothelin (ET) biosynthesis that catalyze the conversion of big ET, the biologically inactive precursor of mature ET. Two enzymes, termed ECE-1 and ECE-2, have been molecularly identified. ECE2(Endothelin-converting enzyme 2) is also named as KIAA0604 and belongs to the methyltransferase superfamily in the N-terminal section, belongs to the peptidase M13 family in the C-terminal section. It is able to process endoproteolytically peptides intermediates to generate biologically active peptides.

Western Blot: Mouse Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: ECE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR