Anti-SYT17 Rabbit Polyclonal Antibody

Supplier: Proteintech
15413-1-AP-150UL
10095-554EA 402 USD
10095-554
Anti-SYT17 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: Mouse Brain Tissue, 1:200-1:2000; FC: HepG2 Cells, N/A

Type: Primary
Antigen: SYT17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR