Anti-USP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15404-1-AP-150UL
10096-848EA 402 USD
10096-848
Anti-USP2 Rabbit Polyclonal Antibody
Antibodies
Ubiquitin, a highly conserved protein involved in the regulation of intracellular protein breakdown, cell cycle regulation, and stress response, is released from degraded proteins by disassembly of the polyubiquitin chains. The disassembly process is mediated by ubiquitin-specific proteases (USPs). USP2 is also named as UBP41. USP2 can interact indirectly with multiple clock proteins, so it remains possible that USP2 may modulate the ubiquitination and function of these other proteins. It has 4 isoforms produced by alternative splicing.

Western Blot: Mouse Testis Tissue, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: USP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR