Anti-SEC13 Rabbit Polyclonal Antibody

Supplier: Proteintech
15397-1-AP-150UL
10094-244EA 537.7 USD
10094-244
Anti-SEC13 Rabbit Polyclonal Antibody
Antibodies
SEC13 is a component of the nuclear pore complex (NPC) and the COPII coat. It is required for esicle biogenesis from endoplasmic reticulum during the transport of proteins. Shuttling of SEC13 between the nucleus and the cytoplasm may couple and regulate functions between these two compartments.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Mouse Brain Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: SEC13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR