Anti-COX8A Rabbit Polyclonal Antibody

Supplier: Proteintech
15368-1-AP-150UL
10085-084EA 402 USD
10085-084
Anti-COX8A Rabbit Polyclonal Antibody
Antibodies
COX8A(Cytochrome c oxidase subunit 8A) is also named as COX8, COX8L and belongs to the cytochrome c oxidase VIII family. This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport. The gene encodes a full length protein of 8 kDa with a transit peptide of 25 amino acid.

Western Blot: Mouse Skeletal Muscle Tissue, 1:1000-1:10000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: COX8A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR