Anti-MYL9 Rabbit Polyclonal Antibody

Supplier: Proteintech
15354-1-AP-150UL
10090-722EA 402 USD
10090-722
Anti-MYL9 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Colon Tissue, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:10-1:100

Type: Primary
Antigen: MYL9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR