Anti-AP3S2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15319-1-AP-150UL
10082-976EA 402 USD
10082-976
Anti-AP3S2 Rabbit Polyclonal Antibody
Antibodies
AP3S2 is a 22-kDa protein that belongs to the adaptor complexes small subunit family. AP3S2 is a subunit of the AP-3 complex which is associated with the Golgi region as well as more peripheral structures. Adaptor protein (AP) complexes are cytosolic heterotetramers that mediate the sorting of membrane proteins in the secretory and endocytic pathways. AP-3 complex is composed of two large adaptins (AP3D1 and AP3B1 or AP3B2), a medium adaptin (AP3M1 or AP3M2) and a small adaptin (APS1 or AP3S2).

Western Blot: Human Testis Tissue, 1:1000-1:10000; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: AP3S2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR