Anti-NDUFV2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15301-1-AP-150UL
10090-992EA 402 USD
10090-992
Anti-NDUFV2 Rabbit Polyclonal Antibody
Antibodies
The NDUFV2 gene encodes the 24-kD subunit of the mitochondrial NADH:ubiquinone oxidoreductase (complex I of the respiratory chain). The protein belongs to the complex I 24 kDa subunit family. It is the core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. NDUFV2 constitutes one genetic risk factor for PD, and the mutation may well be a cause of complex I deficiency in this disease.

Western Blot: Mouse Heart Tissue, 1:500-1:5000; IHC: Human prostate Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: NDUFV2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR