Anti-MRPS25 Rabbit Polyclonal Antibody

Supplier: Proteintech
15277-1-AP-150UL
10090-476EA 524.48 USD
10090-476
Anti-MRPS25 Rabbit Polyclonal Antibody
Antibodies
Mitochondrial ribosomal protein S25 (MRPS25) belongs to the ribosomal protein S25/L51 family.It is a component of the mitochondrial ribosome small subunit (28S) which participates the translation of proteins. This antibody specifically recognizes the 18 kd MRPS25 protein.

Western Blot: HepG2 Cells, 1:500-1:5000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: MRPS25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR