Anti-NCK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15247-1-AP-150UL
10090-866EA 402 USD
10090-866
Anti-NCK1 Rabbit Polyclonal Antibody
Antibodies
NCK family adaptor proteins function to couple tyrosine phosphorylation signals to regulate actin cytoskeletal reorganization that leads to cell motility. Cytoplasmic protein NCK1 (or NCK alpha) is an adapter protein which associates with tyrosine-phosphorylated growth factor receptors, such as KDR and PDGFRB, or their cellular substrates. NCK1 binds a number of intracellular proteins and influences various signaling pathways including GTPase-activating protein of Ras (RasGAP), which is responsible for the down-regulation of Ras. NCK1 is recently reported to regulate the UPR, which secondary to obesity impairs glucose homeostasis and insulin actions.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human breast cancer, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: NCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR