Anti-HLA-A Rabbit Polyclonal Antibody

Supplier: Proteintech
15240-1-AP-150UL
10088-290EA 614.88 USD
10088-290
Anti-HLA-A Rabbit Polyclonal Antibody
Antibodies
Human major histocompatibility complex (MHC) antigens, also referred to as human leukocyte antigens (HLA), are encoded by genes located on the short arm of chromosome 6 (6p21.3). There are two classes of HLA antigens: class I (HLA-A, B and C) and class II (HLA-D). This class I molecules are polymorphic membrane glycoproteins composed of a heavy (alpha) chain (44 kDa) which is encoded by a HLA class I gene (HLA-A, B or C), and β2-microglobulin light (beta) chain (12 kDa). They are involved in the presentation of foreign antigens to the immune system.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Stomach cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: HepG2 Cells, 1:200-1:2000; FC: HepG2 Cells, N/A

Type: Primary
Antigen: HLA-A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR