Anti-RPAIN Rabbit Polyclonal Antibody

Supplier: Proteintech
15220-1-AP-150UL
10093-876EA 402 USD
10093-876
Anti-RPAIN Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse ovary Tissue, 1:200-1:1000; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: RPAIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR