Anti-PSMA7 Rabbit Polyclonal Antibody

Supplier: Proteintech
15219-1-AP-150UL
10092-898EA 402 USD
10092-898
Anti-PSMA7 Rabbit Polyclonal Antibody
Antibodies
PSMA7(Proteasome subunit alpha type-7) is also named as HSPC and belongs to the peptidase T1A family. It is an α-type subunit of the 20S proteasome core complex and participates in degrading proteins through ubiquitin-proteasome pathway (UPP) which plays an important role in the regulation of cell proliferation or cell cycle control, transcriptional regulation, immune and stress response, cell differentiation, and apoptosis..

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: PSMA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR