Anti-OGDH Rabbit Polyclonal Antibody

Supplier: Proteintech
15212-1-AP-150UL
10091-556EA 402 USD
10091-556
Anti-OGDH Rabbit Polyclonal Antibody
Antibodies
OGDH(2-oxoglutarate dehydrogenase, mitochondrial) is also named as OGDC-E1and belongs to the alpha-ketoglutarate dehydrogenase family. It is one of the three enzymatic components of 2-oxoglutarate dehydrogenase complex,which catalyzes the overall conversion of 2-oxoglutarate to succinyl-CoA and CO2. This protein has 3 isoforms produced by alternative splicing.

Western Blot: Mouse lung Tissue, 1:1000-1:10000; IHC: Human lung cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: OGDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR