Anti-THG1L Rabbit Polyclonal Antibody

Supplier: Proteintech
15208-1-AP-150UL
10095-880EA 402 USD
10095-880
Anti-THG1L Rabbit Polyclonal Antibody
Antibodies
IHC: Human endometrial cancer, 1:20-1:200

Type: Primary
Antigen: THG1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR