Anti-RPS3 Rabbit Polyclonal Antibody

Supplier: Proteintech
15198-1-AP-150UL
10081-470EA 402 USD
10081-470
Anti-RPS3 Rabbit Polyclonal Antibody
Antibodies
The ribosomal protein S3 is a subunit of 40S ribosomal protein, and has two distinct function. RPS3 acts as a ribosome protein that contributes to the domain of the ribosome where translation is initiated, and involves in repair of UV damage as an endonuclease. Besides binding RNA within the 40S ribosomal subunit, RPS3 participates in transcription, and apoptosis.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: RPS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR