Anti-NLRP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15182-1-AP-150UL
10091-146EA 402 USD
10091-146
Anti-NLRP2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A549 Cells, 1:200-1:2000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: NLRP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR