Anti-CA12 Rabbit Polyclonal Antibody

Supplier: Proteintech
15180-1-AP-150UL
10083-910EA 402 USD
10083-910
Anti-CA12 Rabbit Polyclonal Antibody
Antibodies
CA12 belongs to the alpha-carbonic anhydrase family. It reversible hydration of carbon dioxide. Overexpression of the zinc enzyme CA12 is observed in certain human cancers. The structure reveals a prototypical CA fold; however, two CA12domains associate to form an isologous dimer, an observation that is confirmed by studies of the enzyme in solution.

Western Blot:BxPC-3 Cells, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200; FC: HEK-293 Cells, N/A

Type: Primary
Antigen: CA12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR