Anti-SARS Rabbit Polyclonal Antibody

Supplier: Proteintech
15162-1-AP-150UL
10094-142EA 402 USD
10094-142
Anti-SARS Rabbit Polyclonal Antibody
Antibodies
SARS(Serine--tRNA ligase, cytoplasmic) Catalyzes the attachment of serine to tRNA(Ser). Is also probably able to aminoacylate tRNA(Sec) with serine, to form the misacylated tRNA L-seryl-tRNA(Sec), which will be further converted into selenocysteinyl-tRNA(Sec).The deduced 514-amino acid protein has a calculated molecular mass of about 59 kD.And it can form a dimer of 120KDa.It is detected a band of 65KDa in MEL cell lines.

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: SARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR