Anti-PGM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15161-1-AP-150UL
10092-156EA 402 USD
10092-156
Anti-PGM1 Rabbit Polyclonal Antibody
Antibodies
PGM1(Phosphoglucomutase-1) is also named as glucose phosphomutase 1 and belongs to the phosphohexose mutase family. It catalyzes the transfer of phosphate between the 1 and 6 positions of glucose. In most cell types, PGM1 isozymes predominate, representing about 90% of total PGM activity. One exception is red cells, where PGM2 is a major isozyme. Defects in PGM1 are the cause of glycogen storage disease type 14 (GSD14). It has 2 isoforms produced by alternative splicing.

Western Blot: Mouse Heart Tissue, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: PGM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR