Anti-PSMB2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15154-1-AP-150UL
10091-876EA 537.7 USD
10091-876
Anti-PSMB2 Rabbit Polyclonal Antibody
Antibodies
PSMB2(proteasome subunit beta type-2) is also named as macropain subunit C7-I, proteasome component C7-I, multicatalytic endopeptidase complex subunit C7-I and belongs to the peptidase T1B family. It is involved in an ATP/ubiquitin-dependent non-lysosomal proteolytic pathway. The up-regulation of PSMB2 may indicate the activated neuronal defensive mechanism in VAD(vitamin A depletion) brain regions, which may underlie the VAD-related psychosis behavior.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: PSMB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR