Anti-PIGK Rabbit Polyclonal Antibody

Supplier: Proteintech
15151-1-AP-150UL
10092-252EA 402 USD
10092-252
Anti-PIGK Rabbit Polyclonal Antibody
Antibodies
Western Blot: HT-1080 Cells, 1:500-1:5000; IHC: Human ovary tumor, 1:20-1:200

Type: Primary
Antigen: PIGK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR