Anti-RPAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15138-1-AP-150UL
10093-878EA 402 USD
10093-878
Anti-RPAP1 Rabbit Polyclonal Antibody
Antibodies
RNA polymerase II-associated protein 1 (RPAP1) encodes a nuclear 153-kDa protein, and forms an interface between the RNA polymerase II (RNAPII) enzyme and chaperone/scaffolding protein, suggesting that it is required to connect RNA polymerase II to regulators of protein complex formation. through its interaction with acetylated histones. RPAP1 is required for interaction of the RNA polymerase II complex with acetylated histone H3 and the subsequent DNA transcription.

Western Blot: HeLa Cells, 1:200-1:2000; IP: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: RPAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR