Anti-SIRT5 Rabbit Polyclonal Antibody

Supplier: Proteintech
15122-1-AP-150UL
10094-412EA 402 USD
10094-412
Anti-SIRT5 Rabbit Polyclonal Antibody
Antibodies
SIRT5, a member fo sirtuin family that is a mitochondrial matrix NAD(+)-dependent deacetylase and mono-ADP-ribosyltransferases. Sirtuins play important role in various cellular processed, such as aging, gene silencing and metabolism. SIRT5 mainly locates in . It is an efficient protein lysine desuccinylase and demalonylase. During prolonged fasting, it can activate CPS1, the committed and regulated enzyme of the urea cycle, and contribute to the regulation of blood ammonia levels.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: SIRT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR