Anti-CAPRIN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15112-1-AP-150UL
10084-058EA 537.7 USD
10084-058
Anti-CAPRIN1 Rabbit Polyclonal Antibody
Antibodies
CAPRIN1(Caprin-1) is a cytoplasmic phosphoprotein which may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. Its levels, except in the brain, are tightly correlated with cellular proliferation. Suppression of expression of human Caprin-1 resulted in slowing of the proliferation rate,which suggests its essential for normal cellular proliferation.

Western Blot: HeLa and ND7 Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200; IP: HeLa Cells, 1:200-1:1000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CAPRIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR