Anti-RPS19 Rabbit Polyclonal Antibody

Supplier: Proteintech
15085-1-AP-150UL
10093-948EA 402 USD
10093-948
Anti-RPS19 Rabbit Polyclonal Antibody
Antibodies
The mammalian ribosome consists of 4 RNA species and approximately 80 different proteins. The ribosomal proteins are encoded by complex gene families that include at least 1 active intron-containing gene and multiple processed pseudogenes. The RPS19 protein is a component of the 40S ribosomal subunit . Diamond-Blackfan anemia (DBA) is a congenital erythroid hypoplasia caused by a functional haploinsufficiency of genes encoding for ribosomal proteins. Among these genes, ribosomal protein S19 (RPS19) is mutated most frequently . It also a p53-related ribosomal protein that possibly involves cellular apoptosis through the BAX/p53 pathway .

Western Blot: Human Testis Tissue, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: RPS19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR