Anti-POLDIP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15080-1-AP-150UL
10092-554EA 402 USD
10092-554
Anti-POLDIP2 Rabbit Polyclonal Antibody
Antibodies
POLDIP2(Polymerase delta-interacting protein 2) is also named as PDIP38, POLD4. It is as a unique positive regulator of Nox4 via its association with p22phox and together with Nox4, has profound effects on Rhodependent cytoskeletal remodeling. It is also as a regulator of cell division. It consists of 368 amino acids and has a predicted molecular mass of 42 kDa, with a potential signal peptide cleavage site after the first N-terminal 48 residues, which would result in a protein of 37 kDa. Endogenous human and rat POLDIP2 localized to the nucleus, plasma membrane, and cytoplasm.

Western Blot: HeLa Cells, 1:500-1:5000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: POLDIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR