Anti-HPS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
15077-1-AP-150UL
10088-434EA 402 USD
10088-434
Anti-HPS1 Rabbit Polyclonal Antibody
Antibodies
HPS1, also named as Hermansky-Pudlak syndrome 1 protein, is a 700 amino acid protein, which is a component of multiple cytoplasmic organelles. HPS1 is Apparently crucial for their normal development and function and involved in intracellular protein sorting.HPS1 is involved in the biogenesis of early melanosomes.

Western Blot:L02 Cells, 1:200-1:2000

Type: Primary
Antigen: HPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR