Anti-METTL3 Rabbit Polyclonal Antibody

Supplier: Proteintech
15073-1-AP-150UL
10090-160EA 537.7 USD
10090-160
Anti-METTL3 Rabbit Polyclonal Antibody
Antibodies
METTL3 is a key S-adenosyl-L-methionine-binding subunit, which is component of a complex multicomponent enzyme that catalyzes the methylation of internal adenosine residues in eukaryotic mRNA, forming N6-methyladenosine. It contains 2 putative nuclear localization signals and 2 consensus methylation motifs.

Western Blot: HEK-293 Cells, 1:500-1:5000; IP: HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: METTL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR