Anti-NDUFS3 Rabbit Polyclonal Antibody

Supplier: Proteintech
15066-1-AP-150UL
10090-980EA 402 USD
10090-980
Anti-NDUFS3 Rabbit Polyclonal Antibody
Antibodies
NDUFS3(NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial) is also named as CI-30kD and belongs to the complex I 30 kDa subunit family. It catalyzes the oxidation of NADH, with the concomitant reduction of ubiquinone and ejection of protons out of the mitochondria. NDUFS3 can be used as a mitochondrial matrix control. The full length has a transit peptide with 36 amino acids.

Western Blot: Human Heart Tissue, 1:200-1:1000; IHC: Human Testis Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: NDUFS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR