Anti-TPPP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
15057-1-AP-150UL
10096-240EA 402 USD
10096-240
Anti-TPPP3 Rabbit Polyclonal Antibody
Antibodies
TPPP3 (tubulin polymerization promoting protein 3), also named as TPPP/p20, is a new member of the TPPP family which binds tubulin and induces tubulin polymerization and microtubule bundling.TPPP3 has been shown to play an important role in cell proliferation and may be implicated in tumorigenesis and metastasis (19633818). TPPP3 was also identified as specific marker for the connective tissues that envelope tendons due to its distinct expression in the developing tendon sheath (19235716). This antibody detects the 19-20 kDa TPPP3 in HeLa and PC-3 cells. The non-specific bands between 40 and 60 kDa were also occasionally observed.

Western Blot:PC-3 Cells, 1:200-1:1000

Type: Primary
Antigen: TPPP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR