Anti-SMOX Rabbit Polyclonal Antibody

Supplier: Proteintech
15052-1-AP-150UL
10094-892EA 537.7 USD
10094-892
Anti-SMOX Rabbit Polyclonal Antibody
Antibodies
SMOX(Spermine oxidase) is also named as SMO, PAO-1 and PAOh1. It belongs to the class of polyamine oxidases (PAOs).The flavoprotein SMO, which specifically oxidizes Spm, is one of the key regulatory enzymes involved in polyamine homeostasis in animal cells . It has 6 isoforms produced by alternative splicing.

Western Blot:PC-13 Cells, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: SMOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR