Anti-GPM6A Rabbit Polyclonal Antibody

Supplier: Proteintech
15044-1-AP-150UL
10087-758EA 402 USD
10087-758
Anti-GPM6A Rabbit Polyclonal Antibody
Antibodies
Glycoprotein M6A (GPM6A, or M6A) is known as a multi-pass transmembrane protein and an abundant cell surface protein on neurons in the central nervous system (CNS). M6A belongs to the myelin proteolipid protein family and is involved in neuronal differentiation, including differentiation and migration of neuronal stem cell. M6A is identified as a retinal membrane protein that is strongly expressed at embryonic stages. In mouse adult retina, M6A plays a role in neuronal plasticity and is involved in neurite and filopodia outgrowth, filopodia motility and probably synapse formation via regulation of MAPK and Src signaling pathways.

Western Blot: Mouse retina Tissue, 1:200-1:2000; IP: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: GPM6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR