Anti-PHF17 Rabbit Polyclonal Antibody

Supplier: Proteintech
15032-1-AP-150UL
10092-188EA 402 USD
10092-188
Anti-PHF17 Rabbit Polyclonal Antibody
Antibodies
PHF17 was identified as a protein partner of the von Hippel-Lindau tumor suppressor pVHL. It is one component of the HBO1 complex which has a histone H4-specific acetyltransferase activity, a reduced activity toward histone H3 and is responsible for the bulk of histone H4 acetylation in vivo. It can act as a transcriptional coactivator that promote acetylation of nucleosomal histone H4 by KAT5. In addition, it ubiquitinates both phosphorylated and non-phosphorylated β-catenin and therefore regulates canonical Wnt signaling in both Wnt-off and Wnt-on phases.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: PHF17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR