Anti-CUTC Rabbit Polyclonal Antibody

Supplier: Proteintech
15030-1-AP-150UL
10085-360EA 402 USD
10085-360
Anti-CUTC Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human endometrial cancer, 1:20-1:200

Type: Primary
Antigen: CUTC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR