Anti-RAB3A Rabbit Polyclonal Antibody

Supplier: Proteintech
15029-1-AP-150UL
10093-188EA 402 USD
10093-188
Anti-RAB3A Rabbit Polyclonal Antibody
Antibodies
Rab3A is a small G-protein of the Rab family, that is implicated in vesicle fusion and regulated secretion, particularly in neurotransmitter release. Rab3 subfamily small G proteins (Rab3A, Rab3B, Rab3C, and Rab3D) control the regulated exocytosis in neuronal/secretory cells. Rab3A is the most abundant Rab GTPase in brain, where it is associated with synaptic vesicles. Rab3a is believed to modulate secretion efficiency by stimulating vesicle recruitment to sites of exocytosis and/ or by recruiting regulatory molecules to the docking/ fusion machinery.

Western Blot: Mouse Brain Tissue, 1:1000-1:10000; IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: RAB3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR