Anti-CPSF4 Rabbit Polyclonal Antibody

Supplier: Proteintech
15023-1-AP-150UL
10085-138EA 402 USD
10085-138
Anti-CPSF4 Rabbit Polyclonal Antibody
Antibodies
The mature 3-prime ends of cellular mRNAs are producted by endonucleolytic cleavage of the primary transcripts, followed by polyadenylation of the upstream cleavage products. The cleavage and polyadenylation specificity factor (CPSF) has a essential role in pre-mRNA 3-prime end processing in that it is required in both the cleavage step and subsequent poly(A) addition . CPSF4 is a component of the cleavage and polyadenylation specificity factor (CPSF) complex, whose other componet is CPSF1, CPSF2, CPSF3 and FIP1L1.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human endometrial cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; IP: HeLa Cells, 1:200-1:1000

Type: Primary
Antigen: CPSF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR