Anti-CITED2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15021-1-AP-150UL
10084-610EA 402 USD
10084-610
Anti-CITED2 Rabbit Polyclonal Antibody
Antibodies
CITED2 is a cAMP-responsive element-binding protein (CBP)/p300-transactivator, with an ED(Glu/Asp)-rich C-terminal domain that acts as an important modulator in the development of the heart . It is proposed to be a negative regulator of HIF-1α through its competitive binding to the CH1 domain of CBP/p300, which mediated signaling and to a decrease in HIF-1-responsive genes such as VEGF . In addition, it is also a co-activator of the transcriptional activity of the TFAP2 isoform, which is an extensively expressed nuclear protein with functions in embryogenesis, inflammation, and stress responses.

Western Blot:SH-SY5Y Cells, 1:200-1:2000

Type: Primary
Antigen: CITED2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR