Anti-DNPEP Rabbit Polyclonal Antibody

Supplier: Proteintech
15016-1-AP-150UL
10086-052EA 402 USD
10086-052
Anti-DNPEP Rabbit Polyclonal Antibody
Antibodies
DNPEP, also named as ASPEP and DAP, plays an important role in intracellular protein and peptide metabolism. It catalyzes the hydrolysis of N-terminal aspartyl and glutamyl residues.

Western Blot: HEK-293 Cells, 1:500-1:5000; IP: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: DNPEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR